Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLC2a Mouse, Clone: S58-205, BD Horizon™

Catalog No. BDB565496 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565496 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565496 Supplier BD Biosciences Supplier No. 565496
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLC2a Monoclonal antibody specifically detects MLC2a in Human, Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLC2a
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone S58-205
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MLC-2a; myosin regulatory light chain 7; MYL7; MYL2A; MYLC2A
Host Species Mouse
Immunogen Human MLC2a Recombinant Protein
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.