Learn More
Promega MLCK Kinase Enzyme System, 10microg
Catalog No. PRV4496 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4496 10 μg
1 options
Catalog No. PRV4496 Supplier Promega Supplier No. V4496
Recombinant human MLCK (1425-1776) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag. MLCK or myosin light chain kinase is a muscle member of the immunoglobulin gene superfamily and a calcium/calmodulin-dependent protein kinase.
  • Easily Screen and Profile MLCK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C