Learn More
Promega MLCK Kinase Enzyme System, 1mg
Catalog No. PRV6588 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6588 1 mg
1 options
Catalog No. PRV6588 Supplier Promega Supplier No. V6588
Add to Cart
Add to Cart
Request a Quote
Recombinant human MLCK (1425-1776) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag. MLCK or myosin light chain kinase is a muscle member of the immunoglobulin gene superfamily and a calcium/calmodulin-dependent protein kinase.
  • Easily Screen and Profile MLCK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C