Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLCK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA515176
Description
MLCK Polyclonal Antibody for Western Blot, IHC (P)
MLCK (MLCK2), now also known as MYLK3, is of the myosin regulatory light chain kinases which facilitates muscle contraction. MLCK, a member of the Ser/Thr protein kinase family, is responsible for smooth muscle contraction via phosphorylation of a specific serine in the N-terminus of myosin light chains (MLC), an event that facilitates myosin interaction with actin filaments. It is a central determinant in the development of vascular permeability and tissue edema formation. In the nervous system it has been shown to control the growth initiation of astrocytic processes in culture and to participate in transmitter release at synapses formed between cultured sympathetic ganglion cells. MLCK acts as a critical participant in signaling sequences that result in fibroblast apoptosis. Smooth muscle and non-muscle isozymes are expressed in a wide variety of adult and fetal tissues and in cultured endothelium with qualitative expression appearing to be neither tissue- nor development-specific. Non-muscle isoform 2 is the dominant splice variant expressed in various tissues. The Telokin isoform, which binds calmodulin, has been found in a wide variety of adult and fetal tissues. MLCK is probably down-regulated by phosphorylation. The protein contains 1 fibronectin type III domain and 9 immunoglobulin-like C2-type domains.
Specifications
| MLCK | |
| Polyclonal | |
| Unconjugated | |
| MYLK3 | |
| caMLCK; cardiac MLCK; cardiac-MLCK; cardiac-MyBP-C associated Ca/CaM kinase; cardiac-MyBP-C-associated Ca/CaM kinase; cardiac-specific myosin light chain kinase; D830007F02Rik; MLC kinase; MLCK; MLCK2; Mylk3; Myosin light chain kinase 3; putative myosin light chain kinase 3; putative myosin light chain kinase 3; myosin light chain kinase 3; RGD1305801 | |
| Rabbit | |
| Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 213435, 91807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q32MK0, Q3UIZ8 | |
| MYLK3 | |
| KLH conjugated synthetic peptide between 40-69 amino acids from the N-terminal region of human MYLK3. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction