Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLCK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576982 Supplier Invitrogen™ Supplier No. PA576982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE).

MLCK (MLCK2), now also known as MYLK3, is of the myosin regulatory light chain kinases which facilitates muscle contraction. MLCK, a member of the Ser/Thr protein kinase family, is responsible for smooth muscle contraction via phosphorylation of a specific serine in the N-terminus of myosin light chains (MLC), an event that facilitates myosin interaction with actin filaments. It is a central determinant in the development of vascular permeability and tissue edema formation. In the nervous system it has been shown to control the growth initiation of astrocytic processes in culture and to participate in transmitter release at synapses formed between cultured sympathetic ganglion cells. MLCK acts as a critical participant in signaling sequences that result in fibroblast apoptosis. Smooth muscle and non-muscle isozymes are expressed in a wide variety of adult and fetal tissues and in cultured endothelium with qualitative expression appearing to be neither tissue- nor development-specific. Non-muscle isoform 2 is the dominant splice variant expressed in various tissues. The Telokin isoform, which binds calmodulin, has been found in a wide variety of adult and fetal tissues. MLCK is probably down-regulated by phosphorylation. The protein contains 1 fibronectin type III domain and 9 immunoglobulin-like C2-type domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLCK
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MYLK3
Gene Accession No. Q32MK0, Q3UIZ8
Gene Alias caMLCK; cardiac MLCK; cardiac-MLCK; cardiac-MyBP-C associated Ca/CaM kinase; cardiac-MyBP-C-associated Ca/CaM kinase; cardiac-specific myosin light chain kinase; D830007F02Rik; MLC kinase; MLCK; MLCK2; Mylk3; Myosin light chain kinase 3; putative myosin light chain kinase 3; putative myosin light chain kinase 3; myosin light chain kinase 3; RGD1305801
Gene Symbols MYLK3
Host Species Rabbit
Immunogen Recombinant full length Human MLCK.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213435, 91807
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.