Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLEC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532923
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bovine, Dog, Bat, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Platypus (100%).
Carbohydrate-binding protein with a strong ligand preference for Glc2-N-glycan. May play a role in the early steps of protein N-glycosylation.
Specifications
MLEC | |
Polyclonal | |
Unconjugated | |
Mlec | |
2410014A08Rik; AI850530; ESTM19; Kiaa0152; malectin; mKIAA0152; Mlec; oligosaccharyltransferase complex subunit (non-catalytic); RGD1307736; X83352 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100050056, 100156249, 100347688, 100755466, 515309, 609274, 9761 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q14165 | |
Mlec | |
Synthetic 18 amino acid peptide from internal region of human MLEC. | |
50 μg | |
Primary | |
Human, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction