Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLEC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532924
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532924 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532924 Supplier Invitrogen™ Supplier No. PA532924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MLEC Polyclonal Antibody for IHC (P)

Carbohydrate-binding protein with a strong ligand preference for Glc2-N-glycan. May play a role in the early steps of protein N-glycosylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLEC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Mlec
Gene Accession No. P02606, Q14165, Q5FVQ4, Q6ZQI3
Gene Alias 2410014A08Rik; AI850530; ESTM19; Kiaa0152; malectin; mKIAA0152; Mlec; oligosaccharyltransferase complex subunit (non-catalytic); RGD1307736; X83352
Gene Symbols Mlec
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from internal region of human MLEC.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050056, 100156249, 100347688, 109154, 304543, 416971, 515309, 609274, 9761
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Chicken, Frog, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.