Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLF1 Interacting Protein Antibody - N-terminal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11085541
Description
MLF1 Interacting Protein Polyclonal specifically detects MLF1 Interacting Protein in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MLF1 Interacting Protein | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
CENP-50, CENP-U, CENPU50, CENPUMLF1-interacting protein, centromere protein of 50 kDa, centromere protein U, FLJ23468, ICEN24, Interphase centromere complex protein 24, KLIP1CENP50, KSHV latent nuclear antigen interacting protein 1, KSHV latent nuclear antigen-interacting protein 1, MLF1 interacting protein, PBIP1, Polo-box-interacting protein 1 | |
Rabbit | |
47.52 kDa | |
0.1 mg | |
Primary | |
This is directed against human MLF1IP protein. A BLAST analysis was used to suggest cross-reactivity with MLF1IP protein from human, chimpanzee (98%), horse (72%), bovine (69%), dog (65%), mouse (56%), rat (55%) and chicken (47%) sources based on identity with the immunizing sequence. Reactivity against homologues from other sources is not known. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.88 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:1400000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q71F23 | |
CENPU | |
This affinity purified MLF1 Interacting Protein Antibody was prepared from whole rabbit serum produced by repeated immunizations with a 200 residue recombinant protein corresponding to the amino terminal end of human MLF1 Interacting Protein protein. (Uniprot: Q71F23) | |
Affinity Purified | |
RUO | |
79682 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction