Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLF1 Monoclonal Antibody (OTI3A3)
GREENER_CHOICE

Catalog No. PIMA525975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525975 Supplier Invitrogen™ Supplier No. MA525975
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLF1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

SERBP1 may play a role in the regulation of mRNA stability. It binds to the 3'-most 134 nt of the SERPINE1/PAI1 mRNA, a region which confers cyclic nucleotide regulation of message decay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3A3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MLF1
Gene Accession No. P58340
Gene Alias Hematopoietic lineage switch 7; HLS7; Mlf1; MLF-1; Myelodysplasia-myeloid leukemia factor 1; myeloid leukemia factor 1; testis tissue sperm-binding protein Li 49e; Unknown (protein for MGC:133499)
Gene Symbols MLF1
Host Species Mouse
Immunogen Full length human recombinant protein of MLF1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4291
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.