Learn More
MLF1IP / PBIP1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | MLF1IP / PBIP1 |
---|---|
Concentration | 1.1 mg/mL |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1954
![]() ![]() |
Rockland Immunochemicals
600401A91 |
100 μg |
Each for $636.24
|
|
|||||
50-199-1955
![]() ![]() |
Rockland Immunochemicals
600401A91S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human MLF1IP protein. A BLAST analysis was used to suggest cross-reactivity with MLF1IP protein from human, chimpanzee (98%), horse (72%), bovine (69%), dog (65%), mouse (56%), rat (55%) and chicken (47%) sources based on homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
The function of this protein remains unknown.Specifications
MLF1IP / PBIP1 | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q71F23 | |
79682 | |
Primary | |
Antibody |
1.1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
1700029A22Rik; CENP50; CENP-50; Cenpu; CENP-U; CENPU50; Centromere protein of 50 kDa; Centromere protein U; ICEN24; Interphase centromere complex protein 24; KLIP1; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; MLF1 interacting protein; MLF1-interacting protein; Mlf1ip; myeloid leukemia factor 1 interacting protein; PBIP1; Polo-box-interacting protein 1 | |
Cenpu | |
-20°C, Avoid Freeze/Thaw Cycles | |
Cenpu |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.