Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLF2 Antibody (OTI1E3), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272721G
Description
MLF2 Monoclonal specifically detects MLF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MLF2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MLF2 | |
Full length recombinant protein of human MLF2 (NP_005430) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E3 | |
Western Blot, Immunohistochemistry | |
Myelodysplasia-myeloid leukemia factor 2, myeloid leukemia factor 2, NTN4 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
8079.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction