Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLF2 Monoclonal Antibody (OTI1E3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525991
Description
MLF2 Monoclonal Antibody for Western Blot, IHC (P)
MLF2 is a protein coding gene.
Specifications
MLF2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15773 | |
MLF2 | |
Full length human recombinant protein of MLF2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1E3 | |
Unconjugated | |
MLF2 | |
AI256856; AU018944; C77284; MLF2; Myelodysplasia-myeloid leukemia factor 2; myeloid leukemia factor 2; myeloid leukemia factor 2 homolog; NTN4 | |
Mouse | |
Affinity chromatography | |
RUO | |
8079 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction