Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLF2 Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
SDP

Catalog No. 502540084
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0084 30 μL
50-167-5441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0084 Supplier OriGene Supplier No. TA504837S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLF2 Monoclonal antibody specifically detects MLF2 in Canine, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MLF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H2
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Acbd3
Gene Accession No. Q7TNY6, Q9H3P7
Gene Alias AI256856; AU018944; C77284; MLF2; Myelodysplasia-myeloid leukemia factor 2; myeloid leukemia factor 2; myeloid leukemia factor 2 homolog; NTN4
Host Species Mouse
Immunogen Full length human recombinant protein of human ACBD3 produced in HKE293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289312, 611888, 64746
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.