Learn More
MLF2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304678A

Description
The recommended shelf life for this product is 1 year from date of receipt.
MLF2 is a protein coding gene.Specifications
MLF2 | |
Polyclonal | |
Unconjugated | |
MLF2 | |
myelodysplasia-myeloid leukemia factor 2, NTN4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30853, 8079 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q15773, Q99KX1 | |
MLF2 | |
Between 175 and 225 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.