Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542166 Supplier Invitrogen™ Supplier No. PA542166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SMQVLQQVDN KFIACLMSTK TEENGEADSY EKQQAQGSGR KKLLSSTLIP Sequence homology: Human: 100%.

This gene is a member of the MutL-homolog (MLH) family of DNA mismatch repair (MMR) genes. MLH genes are implicated in maintaining genomic integrity during DNA replication and after meiotic recombination. The protein encoded by this gene functions as a heterodimer with other family members. Somatic mutations in this gene frequently occur in tumors exhibiting microsatellite instability, and germline mutations have been linked to hereditary nonpolyposis colorectal cancer type 7 (HNPCC7). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLH3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MLH3
Gene Accession No. Q9UHC1
Gene Alias AV125803; BB126472; CAE; CAE1; cell surface glycoprotein; connexin 50; connexin-50; CTRCT1; CX50; CZP1; DNA mismatch repair protein Mlh3; gap junction alpha 8; gap junction alpha-8 protein; gap junction membrane channel protein alpha-8; gap junction protein alpha 8; gap junction protein alpha 8 50kDa; GJA8; HNPCC7; lens fiber protein MP70; lens intrinsic membrane protein MP70; MGC138372; MLH3; MP70; mutL homolog 3; mutL homolog 3 (E coli); MutL protein homolog 3
Gene Symbols MLH3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MLH3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27030
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.