Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MLi-2
GREENER_CHOICESDP

Catalog No. 575650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-565-0 50 mg
57-561-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-565-0 Supplier Tocris Bioscience™ Supplier No. 5756/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective LRRK2 inhibitor

MLi-2 is a potent and selective LRRK2 inhibitor (IC50 = 0.76-3.4 nM depending on assay), which exhibits >295-fold selectivity for over 300 kinases and a diverse panel of receptors and ion channels. MLi-2 is centrally bioavailable and active in vivo.

Specifications

Chemical Name or Material rel-3-[6-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-4-pyrimidyl]-5-[(1-methylcyclopropyl)oxy]-1H-indazole
CAS 1627091-47-7
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C21H25N5O2
Formula Weight 379.46
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.