Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572759
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572759 Supplier Invitrogen™ Supplier No. PA572759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MLIP Polyclonal Antibody for Western Blot, IHC (P), ELISA

Required for precocious cardiac adaptation to stress through integrated regulation of the AKT/mTOR pathways and FOXO1. Regulates cardiac homeostasis and plays an important role in protection against cardiac hypertrophy. Acts as a transcriptional cofactor, represses transactivator activity of ISL1 and MYOCD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLIP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MLIP
Gene Accession No. Q5VWP3
Gene Alias C23H6ORF142; C6orf142; Cardiac Isl1-interacting protein; Cip; flocculation protein FLO11-like; MLIP; muscle-enriched A-type lamin interacting protein; muscular LMNA interacting protein; muscular LMNA-interacting protein; Muscular-enriched A-type laminin-interacting protein; Unknown (protein for MGC:128577)
Gene Symbols MLIP
Host Species Rabbit
Immunogen MLIP antibody for a 16 amino acid synthetic peptide near the carboxy terminus of human MLIP. The immunogen is within the last 50 amino acids of MLIP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90523
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.