Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145170 Supplier Invitrogen™ Supplier No. PA5145170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKALDEPAKT ESVSKDNTLE PPVELYFPAQ LRQQTEELCA TIDKVLQDSL SMHSSDSPSR SPKTLLGSDT Antigen sequence identity: Mouse 81%; Rat 41%.

Required for precocious cardiac adaptation to stress through integrated regulation of the AKT/mTOR pathways and FOXO1. Regulates cardiac homeostasis and plays an important role in protection against cardiac hypertrophy. Acts as a transcriptional cofactor, represses transactivator activity of ISL1 and MYOCD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLIP
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MLIP
Gene Accession No. Q5VWP3
Gene Alias C23H6ORF142; C6orf142; Cardiac Isl1-interacting protein; Cip; flocculation protein FLO11-like; MLIP; muscle-enriched A-type lamin interacting protein; muscular LMNA interacting protein; muscular LMNA-interacting protein; Muscular-enriched A-type laminin-interacting protein; Unknown (protein for MGC:128577)
Gene Symbols MLIP
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 762-831. Recombinant protein control fragment (Product #a href=.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90523
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.