Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570961 Supplier Thermo Scientific Supplier No. PA570961
Add to Cart
Add to Cart

MLIP Polyclonal Antibody

MLIP Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MLIP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MLIP
Gene Accession No. Q5VWP3
Gene Alias C23H6ORF142; C6orf142; Cardiac Isl1-interacting protein; Cip; flocculation protein FLO11-like; MLIP; muscle-enriched A-type lamin interacting protein; muscular LMNA interacting protein; muscular LMNA-interacting protein; Muscular-enriched A-type laminin-interacting protein; Unknown (protein for MGC:128577)
Gene Symbols MLIP
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MLIP.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90523
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.