Learn More
Promega MLK1 Kinase Enzyme System, 1mg
Catalog No. PRV6449 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6449 1 mg
1 options
Catalog No. PRV6449 Supplier Promega Supplier No. V6449
Add to Cart
Add to Cart
Request a Quote
Recombinant human MLK1 (1-433) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Mixed-Lineage Kinase 1 is a mitogen-activated protein kinase kinase kinase capable of activating the c-Jun NH(2)-terminal kinase (JNK) pathway.
  • Easily Screen and Profile MLK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C