Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533916
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533916 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533916 Supplier Invitrogen™ Supplier No. PA533916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Panda, Dog, Horse, Rabbit (100%) Hamster, Elephant, Bovine, Bat, Pig (94%).

MAP3K9 (MLK1) is a serine/threonine protein kinase that functions in the MAP kinase pathway. Plays an important role in the cascades of cellular responses evoked by changes in the environment. Once activated, acts as an upstream activator of the MKK/JNK signal transduction cascade through the phosphorylation of MAP2K4/MKK4 and MAP2K7/MKK7 which in turn activate the JNKs. The MKK/JNK signaling pathway regulates stress response via activator protein-1 (JUN) and GATA4 transcription factors. Plays also a role in mitochondrial death signaling pathway, including the release cytochrome c, leading to apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MAP3K9
Gene Accession No. P80192, Q3U1V8
Gene Alias E130314H24Rik; Map3k9; MEKK9; mitogen activated protein kinase kinase kinase 9; mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity); mixed-lineage kinase 1; MLK1; Prke1; RGD1562149
Gene Symbols MAP3K9
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human MAP3K9.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100054289, 100341731, 338372, 4293, 490765
Target Species Human, Mouse, Canine, Rabbit, Monkey, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.