Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MLK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110619
Description
Immunogen sequence: EGSPQRREKA NGLSTPSESP HFHLGLKSLV DGYKQWSSSA PNLVKGPRSS PALPGFTSLM EMEDEDSEGP GSGESRLQHS PSQSYLCIPF PRGEDGDGPS SDGIHEEPTP VNSATSTPQL TPTNSLKRGG.
MAP3K9 (MLK1) is a serine/threonine protein kinase that functions in the MAP kinase pathway. Plays an important role in the cascades of cellular responses evoked by changes in the environment. Once activated, acts as an upstream activator of the MKK/JNK signal transduction cascade through the phosphorylation of MAP2K4/MKK4 and MAP2K7/MKK7 which in turn activate the JNKs. The MKK/JNK signaling pathway regulates stress response via activator protein-1 (JUN) and GATA4 transcription factors. Plays also a role in mitochondrial death signaling pathway, including the release cytochrome c, leading to apoptosis.
Specifications
| MLK1 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K9 | |
| E130314H24Rik; Map3k9; MEKK9; mitogen activated protein kinase kinase kinase 9; mitogen-activated protein kinase kinase kinase 9; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity); mixed-lineage kinase 1; MLK1; Prke1; RGD1562149 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4293 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P80192 | |
| MAP3K9 | |
| Recombinant Protein corresonding to Human MLK1. Recombinant protein control fragment (Product #RP-88979). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction