Learn More
Promega MLK2 Kinase Enzyme System, 10microg
Catalog No. PRV4476 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4476 10 μg
1 options
Catalog No. PRV4476 Supplier Promega Supplier No. V4476
Recombinant human MLK2 (1-446) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MLK2 or mixed-lineage kinase 2 is a member of the serine/threonine kinase family, which activates MAPK8/JNK1 and MKK4/SEK1.
  • Easily Screen and Profile MLK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C