Learn More
Promega MLK2 Kinase Enzyme System, 1mg
Catalog No. PRV6450 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6450 1 mg
1 options
Catalog No. PRV6450 Supplier Promega Supplier No. V6450
Add to Cart
Add to Cart
Request a Quote
Recombinant human MLK2 (1-446) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MLK2 or mixed-lineage kinase 2 is a member of the serine/threonine kinase family, which activates MAPK8/JNK1 and MKK4/SEK1.
  • Easily Screen and Profile MLK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C