Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575848 Supplier Invitrogen™ Supplier No. PA575848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

MAP3K10 (MLK2) is a serine/threonine kinase which has been shown to activate MAPK8/JNK and MKK4/SEK1. This kinase itself can be phoshorylated, and is thus activated by JNK kinases. MAP3K10 functions preferentially on the JNK signaling pathway, and is reported to be involved in nerve growth factor (NGF) induced neuronal apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLK2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP3K10
Gene Accession No. Q02779, Q66L42
Gene Alias BC028668; BC046514; Map3k10; MAP3K9; MEKK10; MEKK9; mitogen activated protein kinase kinase kinase 10; mitogen-activated protein kinase kinase kinase 10; Mixed lineage kinase 2; mixed-lineage kinase 2; MKN28 derived nonreceptor_type; MKN28 derived nonreceptor_type serine/threonine kinase; MKN28 kinase; MLK1; MLK2; MST; PRKE1; protein kinase MST; serine/threonine kinase
Gene Symbols MAP3K10
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human MLK2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269881, 308463, 4294
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.