Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLK3 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306619H
This item is not returnable.
View return policy
Description
MLK3 Polyclonal antibody specifically detects MLK3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MLK3 | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
EC 2.7.11, EC 2.7.11.25, MEKK11, mitogen-activated protein kinase kinase kinase 11, Mixed lineage kinase 3, MLK3MLK-3, PTK1protein-tyrosine kinase PTK1, SH3 domain-containing proline-rich kinase, SPRKMGC17114, Src-homology 3 domain-containing proline-rich kinase | |
Produced in rabbits immunized with E. coli-derived Human MLK3 fragment. | |
0.1 mL | |
Protein Kinase | |
4296 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction