Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLK3 Polyclonal Antibody, HRP, Bioss
SDP

Catalog No. 502281353
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1353 Supplier Bioss Supplier No. BS2992RHRP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MLK3 Polyclonal antibody specifically detects MLK3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MLK3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene MAP3K11
Gene Accession No. Q16584, Q66HA1, Q80XI6
Gene Alias 2610017K16Rik; MAP3K11; MEKK11; MGC17114; mitogen activated protein kinase kinase kinase 11; mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; Mlk3; MLK-3; protein-tyrosine kinase PTK1; PTK1; RHOE; SH3 domain-containing proline-rich kinase; SPRK; Src-homology 3 domain-containing proline-rich kinase
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MLK3
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26403, 309168, 4296
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.