Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLK4 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP141080R
Description
MLK4 Polyclonal antibody specifically detects MLK4 in Human samples. It is validated for Western BlotSpecifications
MLK4 | |
Polyclonal | |
Western Blot | |
dJ862P8.3, EC 2.7.11, EC 2.7.11.25, mitogen-activated protein kinase kinase kinase MLK4, mixed lineage kinase 4, MLK4, RP5-862P8.2 | |
The immunogen for this product maps to a region between residue 650 and 700 of human mixed lineage kinase 4 using the numbering given in entry NP_115811.2 (GeneID 84451). | |
0.1 mL | |
Protein Kinase | |
84451 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction