Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLK4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548666

Catalog No. PA548666


Only null left
Add to Cart

Description

Description

MLK4 Polyclonal Antibody for Western Blot, IHC (P)

MLK4 or mixed lineage kinase 4 belongs to the superfamily of MAP kinase kinases (MAP3K1) which contain both ser/thr and tyr kinases activity in their catalytic domains. The structure of this kinase family incorporates an N-terminal Src homology (SH3) domain, followed by the kinase domain, a leucine zipper region, and a CDC42 /RAC -interactive binding (CRIB) motif, but divergent C-terminal regions. MLK4 is highly expressed in kidney and pancreas. MLK4 is a negative regulator of TLR4 and does not activate JNK1/MAPK8 pathway, p38/MAPK14, nor ERK2/MAPK1 pathways.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MLK4
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q8VDG6
MAP3K21
KLH conjugated synthetic peptide between 566-593 amino acids from the Central region of mouse Mlk4.
400 μL
Primary
Mouse
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAP3K21
BC021891; cDNA sequence BC021891; dJ862P8.3; Kiaa1804; MAP3K21; Mitogen-activated protein kinase kinase kinase 21; Mitogen-activated protein kinase kinase kinase MLK4; mixed lineage kinase 4; mKIAA1804; MLK4; RGD1306091; similar to Mixed lineage kinase 4
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
234878
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.