Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552460 Supplier Invitrogen™ Supplier No. PA552460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNPTPTGATI ISATGASALP LCPSPAPHSH LPREVSPKKH STVHIVPQRR PASLRSRSDL PQAYPQTAVS QLAQTACVVG RPGPHPTQFL AAKERTKSHV PSLLDADVEG QSRDYTVPLC RMRSKTSRPS IYELEKE Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 34%.

MLK4 or mixed lineage kinase 4 belongs to the superfamily of MAP kinase kinases (MAP3K1) which contain both ser/thr and tyr kinases activity in their catalytic domains. The structure of this kinase family incorporates an N-terminal Src homology (SH3) domain, followed by the kinase domain, a leucine zipper region, and a CDC42 /RAC -interactive binding (CRIB) motif, but divergent C-terminal regions. MLK4 is highly expressed in kidney and pancreas. MLK4 is a negative regulator of TLR4 and does not activate JNK1/MAPK8 pathway, p38/MAPK14, nor ERK2/MAPK1 pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLK4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP3K21
Gene Accession No. Q5TCX8
Gene Alias BC021891; cDNA sequence BC021891; dJ862P8.3; Kiaa1804; MAP3K21; Mitogen-activated protein kinase kinase kinase 21; Mitogen-activated protein kinase kinase kinase MLK4; mixed lineage kinase 4; mKIAA1804; MLK4; RGD1306091; similar to Mixed lineage kinase 4
Gene Symbols MAP3K21
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MLK4. Recombinant protein control fragment (Product #RP-89790).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.