Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLK4 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501563018
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3018 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3018 Supplier Bethyl Laboratories, Inc Supplier No. A302610AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Mixed-lineage kinases (MLKs) function as mitogen-activated protein kinase kinase kinases (MAP3Ks), activating the p38 and JNK signaling cascades. Systematic mutational analyses of tyrosine kinases reveal that a minimum of 30% of colorectal cancer contain at least one mutation in the tyrosine kinases; sampling of colorect cancer cells suggest an important role of MLK4 in this process.

Specifications

Antigen MLK4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MAP3K21
Gene Accession No. Q5TCX8
Gene Alias Mitogen-activated protein kinase kinase kinase; Mixed lineage kinase 4; KIAA1804
Gene Symbols MAP3K21
Host Species Rabbit
Immunogen Between 986 and 1036
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84451
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.