Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLKL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$350.50 - $567.50
Specifications
Antigen | MLKL |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MLKL Polyclonal antibody specifically detects MLKL in Human samples. It is validated for ImmunofluorescenceSpecifications
MLKL | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Necroptosis, Protein Kinase | |
PBS, pH 7.2, 40% glycerol | |
197259 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LQDQGKRSVPSEKLTTAMNRFKAALEEANGEIEKFSNRSNICRFLTASQDKILFKDVNRKLSDVWKELSLLLQVEQRMPVSPISQGASWAQEDQQDADEDRRAFQMLRRDNEKIEASLRRL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title