Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLKL, p Ser345 Antibody (SR1962), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | MLKL |
---|---|
Clone | SR1962 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
MLKL Monoclonal antibody specifically detects MLKL in Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
MLKL | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
A synthesized peptide derived from human phospho-MLKL [Ser345] (Uniprot #: Q9D2Y4) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1962 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Necroptosis, Protein Kinase | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
197259 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title