Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLKL, p Thr357 Antibody (954724), Biotin, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB91871B
Description
MLKL Monoclonal specifically detects MLKL in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow Cytometry (Negative).Specifications
MLKL (p Thr357) | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MLKL | |
Synthetic peptide from human MLKL phosphorylated at T357 [UniProt Q8NB16] | |
100 μL | |
Protein Kinase | |
197259 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry | |
954724 | |
Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow Cytometry (Negative), Immunocytochemistry | |
FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction