Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLKL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543960 Supplier Invitrogen™ Supplier No. PA543960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DVWKELSLLL QVEQRMPVSP ISQGASWAQE DQQDADEDRR AFQMLRRDNE Sequence homology: Cow: 85%; Horse: 91%; Human: 100%; Rat: 90%.

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLKL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Mlkl
Gene Accession No. Q8NB16
Gene Alias 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Gene Symbols Mlkl
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MLKL (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197259
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.