Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLKL Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551001

Catalog No. PA551001


Only null left
Add to Cart

Description

Description

MLKL Polyclonal Antibody detects endogenous levels of MLKL protein.

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MLKL
Polyclonal
Unconjugated
Mlkl
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Rabbit
Antigen affinity chromatography
RUO
197259, 690743, 74568
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NB16, Q9D2Y4
Mlkl
Synthesized peptide derived from the N-terminal region of human MLKL.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.