Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLKL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571886
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571886 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571886 Supplier Invitrogen™ Supplier No. PA571886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MLKL Polyclonal Antibody for Western Blot, IHC (P)

The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLKL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Mlkl
Gene Accession No. Q9D2Y4
Gene Alias 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like
Gene Symbols Mlkl
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 444-472 amino acids from the C-terminal region of mouse Mlkl.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74568
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.