Learn More
Invitrogen™ MLKL Polyclonal Antibody

Description
Antibody detects endogenous levels of total MLKL.
Specifications
Specifications
| Antigen | MLKL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | Mlkl |
| Gene Accession No. | Q8NB16, Q9D2Y4 |
| Gene Alias | 9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like |
| Gene Symbols | Mlkl |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.