Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLKL Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7248416 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168950 20 μg
NB168949 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168950 Supplier Novus Biologicals Supplier No. NBP31693520UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MLKL Polyclonal antibody specifically detects MLKL in Human, Rat samples. It is validated for Western Blot

Specifications

Antigen MLKL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around S441 of mouse MLKL (NP_001297542.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Necroptosis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 197259
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.