Learn More
Invitrogen™ MLKL Recombinant Rabbit Monoclonal Antibody (16H4L15)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ 703737

Description
This antibody is predicted to react with Mouse, Rat, Bovine.
The protein kinase domain is predicted to be catalytically inactive. Molecular function: protein binding. There are two isoforms.
Specifications
MLKL | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8NB16 | |
Mlkl | |
Protein corresponding to human MLKL [aa1-aa154]. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
16H4L15 | |
Unconjugated | |
Mlkl | |
9130019I15Rik; hMLKL; mixed lineage kinase domain like pseudokinase; mixed lineage kinase domain-like; mixed lineage kinase domain-like protein; Mlkl; rCG51567-like | |
Rabbit | |
Protein A | |
RUO | |
197259 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.