Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLL-3 Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE1851
Description
Anti-MLL3 is a highly specific rabbit polyclonal antibody that targets Histone-lysine N-methyltransferase 2C and has been tested in Chromatin Immunoprecipitation (ChIP), Immunohistochemistry (Paraffin), and Western Blotting.
Specifications
MLL-3 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This polyclonal antiody detects MLL-3 protein in human, mouse and rat cells. It targets and epitope within 18 amino acids from the internal region of N-terminal half. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG |
ChIP Assay, Western Blot, Immunohistochemistry (Paraffin) | |
Please refer to lot specific datasheet. | |
Kmt2C,Mll3 | |
A linear peptide corresponding to 18 amino acids from the N-terminal region of human MLL-3. | |
100 μg | |
Epigenetics and Nuclear Function | |
NP_001074852 | |
Mouse, Rat, Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction