Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLL / HRX Monoclonal Antibody (N4.4), Active Motif™
SDP

Catalog No. 501992887
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2887 100 μg
50-199-2888 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2887 Supplier Active Motif Supplier No. 39829
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.

Specifications

Antigen MLL / HRX
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone N4.4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene KMT2A
Gene Accession No. P55200, Q03164
Gene Alias ALL1; ALL-1; CDK6/MLL fusion protein; C-terminal cleavage product of 180 kDa; CXXC7; CXXC-type zinc finger protein 7; Histone-lysine N-methyltransferase 2A; HRX; HTRX; HTRX1; KMT2A; lysine (K)-specific methyltransferase 2A; lysine methyltransferase 2A; Lysine N-methyltransferase 2A; mixed lineage leukemia 1; MLL; MLL cleavage product C180; MLL cleavage product N320; MLL/ENL fusion protein; MLL/GAS7; MLL/GAS7 fusion protein; MLL/GMPS fusion protein; MLL/hCDCrel fusion protein; MLL1; MLL1A; MLL-AF4 der(11) fusion protein; MLL-AF9; Myeloid/lymphoid or mixed-lineage leukemia; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); Myeloid/lymphoid or mixed-lineage leukemia protein 1; N-terminal cleavage product of 320 kDa; p180; p320; rearranged MLL protein; TET1-MLL; Trithorax-like protein; TRX1; WDSTS; Zinc finger protein HRX
Gene Symbols KMT2A
Host Species Mouse
Immunogen This MLL / HRX antibody was raised against a recombinant protein corresponding to amino acids 161-356 of mouse MLL.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214162, 4297
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.