Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLL1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501556871
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
Catalog No. Quantity
50-155-6871 20 μL
1 options

Catalog No. 50-155-6871

Supplier: Bethyl Laboratories, Inc A300087AT

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of ′Lys-4′ of histone H3 (H3K4me) complex and acetylation of ′Lys-16′ of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on ′Thr-3′, less activity toward H3 dimethylated on ′Arg-8′ or ′Lys-9′, and has higher activity toward H3 acetylated on ′Lys-9′. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.

Specifications

Antigen MLL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KMT2A
Gene Accession No. Q03164
Gene Alias ALL-1; Trithorax-like protein
Gene Symbols KMT2A
Host Species Rabbit
Immunogen Between 1320 and 1380
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4297
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.