Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518114
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518114 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518114 Supplier Invitrogen™ Supplier No. PA518114
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

This gene encodes a protein which contains multiple domains including a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET domain. The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL family. This gene is ubiquitously expressed in adult tissues. It is also amplified in solid tumor cell lines, and may be involved in human cancer. Two alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene, however, the full length nature of the shorter transcript is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLL4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene KMT2B
Gene Accession No. Q9UMN6
Gene Alias 2610014H22Rik; ALL1-related protein; ALR; BC032281; BC058659; C430014K11Rik; CXXC10; histone-lysine N-methyltransferase 2B; histone-lysine N-methyltransferase 2B-like; histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; histone-lysine N-methyltransferase MLL4; HRX2; KIAA0304; Kmt2b; Kmt2d; LOC102550344; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase 2B; lysine (K)-specific methyltransferase 2B; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2B; lysine N-methyltransferase 2B; lysine N-methyltransferase 2D; mixed lineage leukemia 2; mixed lineage leukemia gene homolog 2; mKIAA0304; MLL1B; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila) 4; myeloid/lymphoid or mixed-lineage leukemia 2; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia protein 2; myeloid/lymphoid or mixed-lineage leukemia protein 4; Myeloid/lymphoid or mixed-lineage leukemia protein 4 homolog; trithorax homolog 2; trithorax homologue 2; TRX2; Wbp7; WBP-7; WW domain binding protein 7; WW domain-binding protein 7
Gene Symbols KMT2B
Host Species Goat
Immunogen Synthetic peptide sequence (CNCGAKRCRRFLN) corresponding to the C-terminus amino acids of MLL4 (aa 2703-2715).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9757
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.