Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLL4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA540071

Catalog No. PA540071


Only null left
Add to Cart

Description

Description

MLL4 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a protein which contains multiple domains including a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET domain. The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL family. This gene is ubiquitously expressed in adult tissues. It is also amplified in solid tumor cell lines, and may be involved in human cancer. Two alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene, however, the full length nature of the shorter transcript is not known.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MLL4
Polyclonal
Unconjugated
KMT2B
2610014H22Rik; ALL1-related protein; ALR; BC032281; BC058659; C430014K11Rik; CXXC10; histone-lysine N-methyltransferase 2B; histone-lysine N-methyltransferase 2B-like; histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; histone-lysine N-methyltransferase MLL4; HRX2; KIAA0304; Kmt2b; Kmt2d; LOC102550344; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase 2B; lysine (K)-specific methyltransferase 2B; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2B; lysine N-methyltransferase 2B; lysine N-methyltransferase 2D; mixed lineage leukemia 2; mixed lineage leukemia gene homolog 2; mKIAA0304; MLL1B; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila) 4; myeloid/lymphoid or mixed-lineage leukemia 2; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia protein 2; myeloid/lymphoid or mixed-lineage leukemia protein 4; Myeloid/lymphoid or mixed-lineage leukemia protein 4 homolog; trithorax homolog 2; trithorax homologue 2; TRX2; Wbp7; WBP-7; WW domain binding protein 7; WW domain-binding protein 7
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
ELISA, Western Blot
Conc. Not Determined
Whole serum with 0.05% sodium azide
O08550
KMT2B
Three KLH-conjugated synthetic peptides containing an amino acid sequence from the central part of the protein corresponding to mouse Mll4 (Myeloid/lymphoid or mixed-lineage leukemia protein 4).
RUO
75410
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.