Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116906 Supplier Invitrogen™ Supplier No. PA5116906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: THP-1, Mouse testis, Mouse brain Immunogen sequence: GLSDEDYEIL SGLPDSVLYT CGPCAGAAQP RWREALSGAL QGGLRQVLQG LLSSKVVGPL LLCTQCGPDG KQLHPGPCGL QAVSQRFEDG HYKSVHSFME DMVGILMRHS EEGETPDRRA GGQMKGLLLK LLESAFGWFD AHDPKYWRRS TRLPNGVLPN AVLPPSLDHV YAQWRQQEPE TPESGQPPGD PSAAFQGKDP A.

This gene encodes a protein which contains multiple domains including a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET domain. The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL family. This gene is ubiquitously expressed in adult tissues. It is also amplified in solid tumor cell lines, and may be involved in human cancer. Two alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene, however, the full length nature of the shorter transcript is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLL4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KMT2B
Gene Accession No. O08550, Q9UMN6
Gene Alias 2610014H22Rik; ALL1-related protein; ALR; BC032281; BC058659; C430014K11Rik; CXXC10; histone-lysine N-methyltransferase 2B; histone-lysine N-methyltransferase 2B-like; histone-lysine N-methyltransferase 2D; histone-lysine N-methyltransferase MLL2; histone-lysine N-methyltransferase MLL4; HRX2; KIAA0304; Kmt2b; Kmt2d; LOC102550344; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase 2B; lysine (K)-specific methyltransferase 2B; lysine (K)-specific methyltransferase 2D; lysine methyltransferase 2B; lysine N-methyltransferase 2B; lysine N-methyltransferase 2D; mixed lineage leukemia 2; mixed lineage leukemia gene homolog 2; mKIAA0304; MLL1B; MLL2; MLL4; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila) 4; myeloid/lymphoid or mixed-lineage leukemia 2; myeloid/lymphoid or mixed-lineage leukemia 4; myeloid/lymphoid or mixed-lineage leukemia protein 2; myeloid/lymphoid or mixed-lineage leukemia protein 4; Myeloid/lymphoid or mixed-lineage leukemia protein 4 homolog; trithorax homolog 2; trithorax homologue 2; TRX2; Wbp7; WBP-7; WW domain binding protein 7; WW domain-binding protein 7
Gene Symbols KMT2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1370-1570 of human KMT2B (NP_0555421).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102550344, 75410, 9757
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.