Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLLT1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126652AF488
Description
MLLT1 Polyclonal antibody specifically detects MLLT1 in Human samples. It is validated for Western BlotSpecifications
MLLT1 | |
Polyclonal | |
Western Blot | |
ENLENL/MLL fusion, LTG19MLLT1/MLL fusion, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 1, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 1, protein ENL, YEATS domain-containing protein 1, YEATS1myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 1 | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1 using the numbering given in entry NP_005925.2 (GeneID 4298). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4298 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction