Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLLT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583183 Supplier Invitrogen™ Supplier No. PA583183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVMPEGADTV SRPSPDYPML PTIPLSAFSD PKKTKPSHGS KDANKESSKT SKPHKVTKEH RERPRKDSES KSSSKELERE QAKSSKDTSR KLGEGRLPKE EKAPPPKAAF KEPKMALKE Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 84%.

Chromosome band 11q23 is the site of translocations in myeloid and lymphoid acute leukemias, pediatric leukemias, and treatment-induced secondary acute myelogenous leukemia. The translocation breakpoints cluster in a restricted region of the HRX gene resulting in chimeric genes that encode an N-terminal portion of Hrx fused to various partner proteins. Myeloid/lymphoid or mixed-lineage leukemia translocated to 1 (MLLT1) is a nuclear protein with transcriptional transactivation properties that is fused to Hrx in t(11;19) leukemias. The minimal MLLT1 sequence required for transcription activation was narrowed to the C-terminal 90 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLLT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MLLT1
Gene Accession No. Q03111
Gene Alias AA407901; BAM11; CTC-503J8.6; ENL; ENL/MLL fusion; leukemia-associated protein MLLT1; LTG19; MLL/ENL fusion protein; MLLT1; MLLT1 super elongation complex subunit; MLLT1, super elongation complex subunit; MLLT1/MLL fusion; myeloid/lymphoid or mixed lineage leukemia translocation to 1 homolog; myeloid/lymphoid or mixed lineage-leukemia translocation to 1 homolog; myeloid/lymphoid or mixed lineage-leukemia translocation to 4 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocated to, 1; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 1; protein ENL; trithorax; YEATS domain-containing protein 1; YEATS1
Gene Symbols MLLT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MLLT1. Recombinant protein control fragment (Product #RP-102514).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.