Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MLLT10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575235 Supplier Invitrogen™ Supplier No. PA575235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Translocations affecting chromosome 11q23 involve many partner chromosome regions and occur in various leukemias. The 11q23 gene involved in the translocations is MLL. MLLT10 is the partner gene to MLL1 involved in t(10;11)(p12;q23) translocations. In an analysis of two leukemia patients, the in t(10;11)(p12;q23) translocation fuses MLL1, a SET domain containg histone methyltransferase, to the MLLT10 gene. The MLLT10 gene encodes a predicted 1,027-amino acid protein containing an N-terminal zinc finger and a C-terminal leucine zipper domain. The MLLT10 gene is one of the few MLL partner genes to be independently rearranged with a third gene in leukemia, the CALM gene in the t(10;11)(p12;q14) translocation. Chimeric fusion proteins MLL/AF10 and CALM/AF10 consistently retain the leucine zipper motif of MLLT10. The leucine zipper interacts with GAS41, a protein previously identified as the product of an amplified gene in a glioblastoma. GAS41 interacts with integrase interactor-1 (INI1), a component of the SWI/SNF complex, which acts to remodel chromatin and to modulate transcription. Retention of the leucine zipper in the MLL and CALM fusions suggested that a key feature of these chimeric proteins may be their ability to interfere in normal gene regulation through interaction with the adenosine triphosphate-dependent chromatin remodeling complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLLT10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MLLT10
Gene Accession No. O54826, P55197
Gene Alias Af10; af-10; ALL1-fused gene from chromosome 10 protein; B130021D15Rik; D630001B22Rik; DKFZp686E10210; MGC75086; mKIAA4140; MLLT10; myeloid/lymphoid or mixed lineage-leukemia translocation to 10 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 10; myeloid/lymphoid or mixed-lineage leukemia translocated to, 10; myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10; protein AF-10; translocated to, 10; type I AF10 protein; type III AF10 protein; type IV AF10 protein
Gene Symbols MLLT10
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 200-250 of Human MLLT10.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17354, 361285, 8028
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.