Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MLLT6 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122995AF594
Description
MLLT6 Polyclonal antibody specifically detects MLLT6 in Human, Mouse samples. It is validated for Western BlotSpecifications
MLLT6 | |
Polyclonal | |
Western Blot | |
AF17Myeloid/lymphoid or mixed-lineage leukemia, translocated to, 6, ALL1-fused gene from chromosome 17 protein, FLJ23480, myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog); translocatedto, 6, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 6, protein AF-17, trithorax homolog | |
The immunogen recognized by this antibody maps to a region between residue 1043 and 1093 of human AF17 (MLLT6, myeloid/lymphoid or mixed-lineage leukemia translocated to, 6 ) using the numbering given in entry NP_005928.2 (GeneID 4302). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4302 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction